Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Voriconazole Without Stress on Chromolith Performance 100-3mm
HPLC Analysis of Voriconazole Without Stress on Chromolith Performance 100-3mm
Conditions
column Chromolith® Performance 100 x 3 mm (1.52001.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] water; (50:50, A:B)
flow rate 425 μL/min
pressure 4032 psi (28 bar)
sample Voriconazole 50 μg/mL
injection 5 μL
detector 256 nm cell: 1.4 μL