Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Warfarin™ Anticoagulant on Ascentis® RP-Amide
HPLC Analysis of Warfarin™ Anticoagulant on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565324-U)
column temp. 35 °C
mobile phase [A] 20 mM phosphoric acid (pH 2.0 unadjusted): [B] methanol; (30:70, A:B)
flow rate 1.0 mL/min
sample as indicated in 20 mM phosphoric acid (pH 2.0)
injection 10 µL
detector UV, 280 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Anticoagulants
Featured Industry Clinical
Pharmaceutical (small molecule)
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC, Warfarin is a trademark of Wisconsin Alumni Research Foundation