Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Warfarin™ Enantiomers on Astec® CHIROBIOTIC® V (Method 2)
HPLC Analysis of Warfarin™ Enantiomers on Astec® CHIROBIOTIC® V (Method 2)
Conditions
column CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 µm particles (11024AST)
column temp. ambient
mobile phase [A] 0.1% triethylamine acetate, pH 5.0; [B] THF; (80:20, A:B)
flow rate 1 mL/min
sample 4 mg/mL in methanol
injection 1 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Clinical, Chiral Applications, Clinical, Forensic & Toxicology, anticoagulants, Anticoagulants, Pharmaceutical (small molecule), All Pharmaceutical Applications, Therapeutic Drug Monitoring, Anticoagulants/Antiplatelet Drugs
Featured Industry Clinical
Pharmaceutical (small molecule)
Legal Information Warfarin is a trademark of Wisconsin Alumni Research Foundation, CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC