Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Withania (Aswagandha) Standard on Ascentis® Express F5
HPLC Analysis of Withania (Aswagandha) Standard on Ascentis® Express F5
Conditions
column Ascentis Express F5, 10 cm x 2.1 mm I.D., 2.7 µm particles (53569-U)
column temp. 35 °C
mobile phase [A] water; [B] acetonitrile
gradient 20 to 100% B in 10 min: held at 100% B for 0.5 min
flow rate 0.3 mL/min
pressure 2756 psi (190 bar)
sample 20 µg/mL in 80:20, water:methanol
injection 5 µL
detector UV, 227 nm
Description
Analysis Note This application demonstrates the suitability of Ascentis Express F5 for the efficient separation of withania standards.
Categories Analytical Chromatography, Herbal medicines
Featured Industry Vitamins, Nutraceuticals, and Natural Products