HPLC Analysis of Xiaoyao Wan on Purospher(R) STAR RP-18 endcapped
HPLC Analysis of Xiaoyao Wan on Purospher® STAR RP-18 endcapped
Conditions
column Purospher STAR RP-18 endcapped 150 x 4.6 mm (1.51455.0001)
column temp. 30 °C
mobile phase [A] 0.1% H3PO4 aqueous solution; [B] acetonitrile; (85:15, A:B)
flow rate 1 mL/min
pressure 1537 psi (106 Bar)
sample Dilute ethanol solution (dilute 529 mL ethanol with water to 1000 mL). (60 μg/mL (60 ppm) Paeoniflorin in diluent.)
injection 10 μL
detector 230 nm cell: 1 μL/10mm
Description
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information Purospher is a registered trademark of Merck KGaA, Darmstadt, Germany