Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Xiaoyao Wan on Chromolith(R) HighResolution RP-18 endcapped
HPLC Analysis of Xiaoyao Wan on Chromolith® HighResolution RP-18 endcapped
Conditions
column Purospher STAR RP-18 endcapped 150 x 4.6 mm (1.52022.0001)
column temp. 30 °C
mobile phase [A] 0.1% H3PO4 aqueous solution; [B] acetonitrile; (85:15, A:B)
flow rate 1.0 mL/min
pressure 826 psi (57 bar)
sample Dilute ethanol solution (dilute 529 mL ethanol with water to 1000 mL). (60 μg/mL (60 ppm) Paeoniflorin in diluent.)
injection 10 μL
detector 230 nm cell: 1μL/10mm
Description
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany