HPLC Analysis of Xylose, Fructose, Glucose and Saccharose on Chromolith(R) NH2 100 - 4.6mm
HPLC Analysis of Xylose, Fructose, Glucose and Saccharose on Chromolith® NH2 100 - 4.6mm
Conditions
column Chromolith® NH2 100 x 4.6mm (1.52028.0001)
column temp. 23 °C ambient
mobile phase [A] acetonitrile; [B] water; (75:25, A:B)
flow rate 1.0 mL/min
pressure 160 psi (11 bar)
sample Xylose 2.28 mg/mL, Fructose 6.45 mg/mL, Glucose 9.20 mg /mL, and Saccharose 9.42 mg/mL diluted in mobile phase.
injection 2 μL
detector 190 nm cell: 11 μL
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany