HPLC Determination of Free Undecenoic Acid in Antifungal Powder on Ascentis(R) Express C18
HPLC Determination of Free Undecenoic Acid in Antifungal Powder on Ascentis® Express C18
sample preparation Sonicate samples for 30 minutes in acetonitrile, filter with 33 mm Hydrophobic PTFE, 0.45 μm filter, dilute with mobile phase.
column Ascentis Express C18 2.7 μM 10 cm x 2.1 mm particles (53823-U)
column temp. 30 °C
mobile phase 40/60, [A]/[B]; [A] 95/5 water/acetonitrile in 0.1% phosphoric acid, [B] 5/95 water /acetonitrile in 0.1% phosphoric acid
flow rate 0.3 mL/min
pressure 2608 psi (180 bar)
sample 100 μg/mL antifungal powder extracted in acetonitrile diluted in mobile phase
injection 1 μL
detector 200 nm UV
Analysis Note Undecenoic acid is an eleven-carbon monounsaturated fatty acid and is naturally found in the body. It is used as a topical antimicrobial in the following forms; powder, ointment or spray. It treats a wide variety of skin infections that are caused by fungus; jock itch, athlete’s foot, diaper rash or itching, burning in the groin area. This method demonstrates free undecenoic acid for detection without the use of any derivatization reagents. Acetonitrile is most effective at solvating the free undecenoic acid without dissociating zinc undecylenate and thus provides discrimination based on differential solubility.
Featured Industry Industrial Hygiene