HPLC Determination of Total Undecenoic Acid in Antifungal Powder After the Complete Dissociation of Zinc Undecylenate on Ascentis(R) Express C18
HPLC Determination of Total Undecenoic Acid in Antifungal Powder After the Complete Dissociation of Zinc Undecylenate on Ascentis® Express C18
Conditions
sample preparation Sonicate samples for 30 minutes in 0.1% ammonium hydroxide in methanol, filter with 33 mm Hydrophobic PTFE, 0.45 μm filter, dilute with mobile phase.
column Ascentis Express C18 2.7 μM 10 cm x 2.1 mm particles (53823-U)
column temp. 30 °C
mobile phase 40/60, [A]/[B]; [A] 95/5 water/acetonitrile in 0.1% phosphoric acid, [B] 5/95 water /acetonitrile in 0.1% phosphoric acid
flow rate 0.3 mL/min
pressure 2623 psi (181 bar)
sample 100 μg/mL antifungal powder extracted with 0.1% ammonium hydroxide in methanol diluted in mobile phase
injection 1.0 μL
detector 200 nm UV
Description
Analysis Note Undecenoic acid is an eleven-carbon monounsaturated fatty acid and is naturally found in the body. It is used as a topical antimicrobial in the following forms; powder, ointment or spray. It treats a wide variety of skin infections that are caused by fungus; jock itch, athlete’s foot, diaper rash or itching, burning in the groin area. This method demonstrates the complete dissociation of zinc undecylenate to undecenoic acid for detection without the use of any derivatization reagents. This application along with the free undecenoic acid determined in acetonitrile application may be utilized to determine the amount of zinc undecylenate in the powder formulation.
Featured Industry Industrial Hygiene