Sildenafil Citrate Method (HPLC) following USP Monograph on Ascentis Express C18 with UV detection
Sildenafil, sold under the brand name Viagra, among others, is a medication used to treat erectile dysfunction and pulmonary arterial hypertension. In 1994, Pfizer filed a patent covering the use of sildenafil to treat erectile dysfunction. This patent will expire in 2019.
Ascentis® Express C18 columns can be used for the HPLC methods within the sildenafil citrate USP Monograph, here illustrated with appropriate analytical data to verify that system suitability criteria are met per USP41-NF36 guidelines.
Same experimental conditions are used for both assay and organic impurities (related substances) but in the latter method the chromatograph should run for three (3) times the retention time of sildenafil. The suitability requirements for the assay method are defined reproducibility (not more than 0.85% for six replicates) and peak tailing factor (not more than 1.5) whereas for organic impurities the relative retention times for sildenafil, sildenafil N-oxide, and sildenafil related compound A are about 1.0, 1.2, and 1.7, respectively. The resolution should be not less than 2.5 between sildenafil N-oxide and sildenafil (using the system suitability solution), and the signal-to-noise ratio not less than 10 (using the sensitivity solution).
Sildenafil Citrate
Experimental Conditions
Column Ascentis® Express C18 150x4.6 mm, 5 µm Injection volume 10 µL
Detection UV @ 290 nm (analytical flow cell; 10 µL) Flow rate 1.0 mL/min
Buffer Dilute 7 mL of triethylamine with water to 1 L.
Stir and adjust with phosphoric acid to a
pH of 3.0 ± 0.1.
Temperature 30 °C
Mobile phase Mix buffer, methanol and acetonitrile 58:25:17 (v/v/v) Pressure drop 160 bar (2321 psi)
Diluent Mobile phase    
SST solution Dissolve 70 mg of Sildenafil Citrate in 1 mL of a solution of hydrogen peroxide and anhydrous formic acid (2:1).
Allow to stand for at least 10 min to generate sildenafil N-oxide, and then dilute with mobile phase to 250 mL.
Sample solution 0.7 mg/mL of Sildenafil Citrate in mobile phase.
Diluted sample solution 1.4 µg/mL of Sildenafil Citrate in mobile phase from the sample solution
Sensitivity solution 0.35 µg/mL of Sildenafil Citrate in mobile phase from the diluted sample solution
Chromatographic Data (System Suitability Solution)
  Compound Retention
Time (min)
Resolution Theoretical
Plates
Tailing
Factor
1 Sildenafil Citrate 6.0 - 11,607 1.1
2 N-oxide 7.0 3.7 11,499 1.1
Chromatographic Data (Sensitivity Solution)
  Compound Retention Time (min) Tailing factor Theoretical Plates Signal-to-noise ratio
1 Sildenafil Citrate 6.0 1.0 11,499 17.0
Validation and verification
1.Specificity: Inject solution and determine the retention time of desired analyte in presence of other component like impurities and excipient.
    Retention Time (min) RRT Tailing Factor
1 Sildenafil citrate 6.0 1.0 1.0
2 N-oxide 7.0 1.2 1.2
3 Related compound A 9.5 1.6 1.1
2. Standard Repeatability (0.35ppm)
STD 1 4881
STD 2 4900
STD 3 4797
STD 4 4842
STD 5 4860
Mean 4868
Standard Deviation 40.0
RSD (%) 0.8
3. Linearity  
Concentration (ppm) Area of Sildenafil Citrate
0.35 4783
1.4 20135
3.5 48171
7 94168
14 187369
22.4 299620
28 377224
33.6 461333
42 558476
LOQ 0.2 ppm (S/N 10)
LOD 0.06 ppm (S/N 3.0)