Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS Analysis of 3-Methoxytyramine and Metanephrines on SeQuant ZIC-HILIC
LC/MS Analysis of 3-Methoxytyramine and Metanephrines on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, 10 cm x 2.1 mm, 3.5 µm, 100 Ã… particles (1.50441.0001)
column temp. 25 °C
mobile phase 5 mM ammonium formate with 0.1% formic acid in 85:15, acetonitrile:water
flow rate 0.5 mL/min
pressure 3500 psi (241 bar)
sample 200 ng/mL in acetonitrile
injection 1 µL
detector MS, ESI+, combined SIR
Description
Categories Analytical Chromatography, Catecholamines
Featured Industry Clinical
suitability application for LC-MS