LC/MS Analysis of Acetylcysteine Granules on SeQuant(R) ZIC-HILIC
LC/MS Analysis of Acetylcysteine Granules on SeQuant® ZIC-HILIC
Conditions
column Sequant ZIC-HILIC 250 x 4.6mm, 5μm, 200 Ã… (1.50458.0001)
mobile phase [A] KH2PO4,pH=5.6; [B] Acetonitrile; (85:15, A:B)
flow rate 1.0 mL/min
sample 1mg/ml of Acetylcysteine in Acetonitrile/water solution(70/30) (1g of Acetylcysteine Granules(equals to 100mg of pure Acetylcystein) was dissolved in 30ml of pure water,diluted by acrtonitrile to 100ml. )
injection 1 μL
detector 205 nm
Description
Featured Industry Life Science and Biopharma
Legal Information SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany
suitability application for LC-MS