Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS Analysis of Amino Acids on Ascentis® Express HILIC
LC/MS Analysis of Amino Acids on Ascentis® Express HILIC
Conditions
column Ascentis Express HILIC, 15 cm x 4.6 mm I.D., 2.7 µm particles (53981-U)
column temp. 35 °C
mobile phase [A] 13 mM ammonium acetate in 10:90 (v/v) water: acetonitrile; [B] 13 mM ammonium acetate in water
gradient held at 0% B for 1 min; 0 to 90% B in 19 min
flow rate 1.0 mL/min
sample 10 mg/L in Mobile Phase A
injection 10 µL
detector ESI(+)
Description
Categories Analytical Chromatography, Amino Acids