Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS Analysis of Annato (or Roucou or Achiote) on Chromolith® FastGradient RP-18 endcapped
LC/MS Analysis of Annato (or Roucou or Achiote) on Chromolith® FastGradient RP-18 endcapped
Conditions
column Chromolith FastGradient RP-18, 5 cm x 2.0 mm I.D, 2 µm particles (1.52007.0001)
column temp. 25 °C
mobile phase [A] 0.1% formic acid in water; [B] 0.1% formic acid in acetonitrile
gradient 55 to 64% B in 0.5 min, to 70% B in 3 min, to 90% B in 0.5 min, held at 90% B for 2 min, to 55% B in 0.5 min, held at 55% for 2.5 min
flow rate 0.4 mL/min
pressure 562-317 psi (39 - 22 bar)
injection 10 µL
detector ESI-MS(+) (m/z range 378-396), BPC
Description
Categories Analytical Chromatography, Dyes
Featured Industry Food and Beverages
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany
Other Notes Sample Prep: Dissolve 2.2 mg/mL cis-norbixin and 0.12 mg/mL cis-bixin in diluent. After dissolution of the analytes the solution was heated for 5.5 hr at 75 °C under reflux to speed up transformation of cis- to trans-isomers and thereafter diluted 1:10 with acetonitrile and water (50:50 v/v).
suitability application for LC-MS