Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS Analysis of Antifungals on Ascentis® Express Phenyl-Hexyl
LC/MS Analysis of Antifungals on Ascentis® Express Phenyl-Hexyl
Conditions
column Ascentis Express Phenyl-Hexyl, 10 cm x 2.1 mm I.D., 2.7 µm particles (53336-U)
column temp. 35 °C
gradient [A] 10 mM ammonium acetate; [B] acetonitrile
gradient 2% B for 1 min; to 100% B in 10 min; held at 100% B for 1 min
flow rate 0.3 mL/min
pressure 2930 psi (202 bar)
sample Antifungals, 1-10 µg/mL in 99:1, water:methanol
injection 0.5 µL
detector MS, ESI+, XIC
Description
Categories Analytical Chromatography, Antifungals