LC/MS Analysis of Cefazolin on SeQuant(R) ZIC-HILIC
LC/MS Analysis of Cefazolin on SeQuant® ZIC-HILIC
column SeQuant ZIC-cHILIC, PEEK, 150 x 4.6 mm, 3 μm, 100 Ã… (1.50661.0001)
column temp. 30 °C
mobile phase [A] acetonitrile; [B] 7.7 g of ammonium acetate in 1000 ml water (100 mM); (90:10, A:B)
flow rate 1.5 mL/min
pressure 1652 psi 118 Bar
sample 50 ppm cefazolin and 400 ppm salicylic acid in mobile phase in mobile phase
injection 10 μL
detector 254 nm cell: 8 μL
Featured Industry Pharmaceutical (small molecule)
Legal Information SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany
suitability application for LC-MS
Related Links