Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS Analysis of Chondroitin Sulfate on SeQuant ZIC-HILIC
LC/MS Analysis of Chondroitin Sulfate on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC column (250 x 4.6 mm, 5 μm, 200 Ã…) (1.50458.0001)
column temp. 30 °C
mobile phase [A] acetonitrile; [B] ammonium acetate; (60:40, A:B)
flow rate 1.0 mL/min
pressure 1200 psi (80 bar)
injection 10 μL
detector Ahent 1200, ELSD
Description
Featured Industry Vitamins, Nutraceuticals, and Natural Products