LC/MS Analysis of Citicoline on SeQuant(R) ZIC-HILIC
LC/MS Analysis of Citicoline on SeQuant® ZIC-HILIC
Conditions
column SeQuant® ZIC-cHILIC, PEEK, 150 x 4.6 mm, 3 μm, 100 Ã… (1.50661.0001)
column temp. 30 °C
mobile phase [A] acetonitrile; [B] 7.7 g ammonium acetate and dissolve in in 1L MilliQ water; (70:30, A:B)
flow rate 0.75 mL/min
pressure 856 psi (59 bar)
sample Weigh 25 mg citicoline in 50 ml volumetric flask. Dissolve in diluent.
injection 20 μL
detector 276 nm
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany
suitability application for LC-MS