LC/MS Analysis of D-Mannose on SeQuant ZIC-HILIC
LC/MS Analysis of D-Mannose on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-cHILIC, PEEK, 150 x 4.6 mm, 5 μm, 100 Ã… (1.50661.0001)
column temp. 40 °C (oven), 40 °C (detector cell)
mobile phase [A] acetonitrile; [B] 3.54 g of ammonium acetate in 500 ml milliQ water; (80:20, A:B)
flow rate 0.5 mL/min
pressure 421 psi (29 bar)
sample see Description text
injection 10 μL
detector Shimadzu Prominence, R.I.
Description
Analysis Note Pipette out 2.5 ml of suspension into a 50 ml volumetric flask. Add 30 ml of diluent. Sonicate for 10 minutes. Centrifuge the solution at 10000 rpm for 10 minutes. Filter through 0.2 um filter paper.