Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS Analysis of Erythromycin Analogs on Ascentis® Express HILIC
LC/MS Analysis of Erythromycin Analogs on Ascentis® Express HILIC
Conditions
column Ascentis Express HILlC, 15 cm x 4.6 mm I.D., 2.7 µm particles (53981-U)
column temp. 35 °C
mobile phase [A] 100 mM ammonium formate, pH 3.0 (titrated with formic acid); [B] acetonitrile; (10:90, A:B)
flow rate 1.0 mL/min
pressure 1247 psi (86 bar)
injection 5 µL
detector ESI(+); SIR
Description
Analysis Note This application shows the suitability of Ascentis Express Silica columns for the analysis of erythromycin analogs in aqueous normal phase/HILIC mode of chromatography. Other relatively hydrophobic aminoglycosides have also been shown to separate under these conditions.
Categories Analytical Chromatography, Antibiotics, Antimicrobials