LC/MS Analysis of Fructose and NaCl on SeQuant ZIC-HILIC
LC/MS Analysis of Fructose and NaCl on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-cHILIC, PEEK, 150 x 4.6 mm, 5 μm, 100 Ã… (1.50461.0001)
column temp. 40 °C
mobile phase [A] acetonitrile; [B] ammonium acetate 100 mM pH 5; (80:20, A:B)
flow rate 1.5 mL/min
pressure 2000 psi (135 bar)
sample 1000 ppm Fructose & 90ppm NaCl
injection 20 μL
detector Sedere Sedex 85 ELSD (cell: analytical nebulizer)