Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS Analysis of Fructose and NaCl on SeQuant(R) ZIC-HILIC
LC/MS Analysis of Fructose and NaCl on SeQuant® ZIC-HILIC
Conditions
column SeQuant ZIC-cHILIC, PEEK, 150 x 4.6 mm, 5 μm (1.50461.0001)
column temp. 40 °C
mobile phase [A] acetonitrile; [B] 100 mM Ammonium acetate, adjusted with acetic acid to pH 5.0; (80:20, A:B)
flow rate 1.5 mL/min
pressure 2000 psi 135 Bar
sample 1000 ppm Fructose and 90 ppm NaCl (Diluent Acetonitrile and water 80:20 (v/v))
injection 20 μL
detector ELSD (Analytical nebulizer)
Description
Featured Industry Food and Beverages
Legal Information SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany