Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS Analysis of Hydroxyamphetamine Hydrobromide on SeQuant(R) ZIC-HILIC
LC/MS Analysis of Hydroxyamphetamine Hydrobromide on SeQuant® ZIC-HILIC
Conditions
column SeQuant® ZIC-cHILIC, PEEK, 150 x 4.6 mm, 3 μm, 100 Ã… (1.50661.0001)
column temp. 25 °C
mobile phase [A] acetonitrile; [B] 1.361 g of monopotassium phosphate in 1000 ml water (10mM); (70:30, A:B)
flow rate 1.0 mL/min
pressure 1174.5 psi (81 bar)
sample Hydroxyamphetamine Hydrobromide RS 100 ppm
injection 10 μL
detector 230 nm cell: 13 μL
Description
Featured Industry Clinical
Legal Information SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany
suitability application for LC-MS