Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS Analysis of Moniliformin on SeQuant® ZIC-HILIC
LC/MS Analysis of Moniliformin on SeQuant® ZIC-HILIC
Conditions
column Sequant ZIC-HILIC, PEEK, 150 x 4.6 mm, 3.5 μm, 200 Å (1.50449.0001)
mobile phase  [A] acetonitrile; [B] 100 mM ammonium formate
gradient 5 to 15% B in 15 min at 0.5 mL/min; to 50% B in 1 min at 1 mL/min; held at
50% B for 4 min; to 5% B in 1 min; held at 5% B at 0.5 mL/min for 2 min
flow rate 0.5 mL/min
detector diode array at 229 nm, and ESI-MS(-)
injection: 3 μL
Description
Featured Industry Food and Beverages
Legal Information SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany
suitability application for LC-MS