Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS Analysis of N-Acetyl-L-Cysteine and Citrulline on SeQuant® ZIC-HILIC
LC/MS Analysis of N-Acetyl-L-Cysteine and Citrulline on SeQuant® ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 100 x 4.6 mm, 5 μm, 200 Å (1.50453.0001)
mobile phase [A] acetonitrile; [B] ammonium acetate; (70:30, A:B)
flow rate 0.5 mL/min
detector ESI MS, negative ion mode
injection: 10 μL
Description
Featured Industry Life Science and Biopharma
Legal Information SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany
suitability application for LC-MS