LC/MS Analysis of Nadolol on SeQuant(R) ZIC-HILIC
LC/MS Analysis of Nadolol on SeQuant® ZIC-HILIC
Conditions
column SeQuant ZIC-cHILIC, PEEK, 150 x 4.6 mm, 3 μm, 100 Ã… (1.50661.0001)
column temp. 25 °C
mobile phase [A] acetonitrile; [B] 1.927 g of ammonium acetate in 1000 ml water (25 mM); (85:15, A:B)
flow rate 1.0 mL/min
pressure 841 psi (57 bar)
sample Nadolol RS 500 ppm
injection 20 μL
detector 273 nm cell: 13 μL
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany
suitability application for LC-MS