LC/MS Analysis of Oseltamavir (Tamiflu) and Oseltamavir Carboxylate on SeQuant(R) ZIC-HILIC
LC/MS Analysis of Oseltamavir (Tamiflu) and Oseltamavir Carboxylate on SeQuant® ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 20 x 2.1 mm, 3.5 μm, 100 Ã… (1.50439.0001)
mobile phase [A] acetonitrile; [B] ammonium acetate; (85:15, A:B)
flow rate 1.0 mL/min
injection 2.5 μL
detector Agilent 1200 LC-system coupled online to an API5000 triple quadrupole
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany
suitability application for LC-MS