Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS Analysis of Peptides, Hydrophobic, on Ascentis® C18
LC/MS Analysis of Peptides, Hydrophobic, on Ascentis® C18
Conditions
column Ascentis C18, 10 cm x 2.1 mm I.D., 5 µm particles (581326-U)
column temp. ambient
mobile phase [A] 50:50 (v/v) 50 mM formic acid, pH 3.0 with acetic acid:water; [B] 50:50 (v/v) 50 mM formic acid, pH 3.0 with acetic acid:acetonitrile
gradient 20 to 50% B in 15 min
flow rate 0.2 mL/min
sample as indicated in 10 mM formic acid
injection 4 µL
detector ESI (+)
Description
Categories Analytical Chromatography, Peptides