LC/MS Analysis of Quarternary Ammonium Compounds on Ascentis® Si (Silica)
LC/MS Analysis of Quarternary Ammonium Compounds on Ascentis® Si (Silica)
Conditions
column Ascentis Si, 15 cm x 4.6 mm I.D., 5 µm particles (581512-U)
column temp. 35 °C
mobile phase [A] 0.1% ammonium acetate in water (pH unadjusted): [B] 0.1% ammonium acetate in acetontrile; (10:90, A:B)
flow rate 1 mL/min. split to the MS
sample as indicated in 0.1% ammonium acetate in 10:90:water:acetonitrile
injection 5 µL
detector MS, ESI (+) in selected ion recording (SIR) mode
Description
Categories Analytical Chromatography, Anticholinergics
Featured Industry Chemicals and Industrial Polymers