LC/MS Analysis of Ractopamine on SeQuant(R) ZIC-HILIC
LC/MS Analysis of Ractopamine on SeQuant® ZIC-HILIC
Conditions
column SeQuant® ZIC-cHILIC, PEEK, 150 x 4.6 mm, 3 μm, 100 Ã… (1.50661.0001)
column temp. 35 °C
mobile phase [A] acetonitrile; [B] 1.361 g of monopotassium phosphate in 1000 ml water (10mM); (80:20, A:B)
flow rate 1.0 mL/min
pressure 913.5 psi 63 Bar
sample 50 ppb Ractopamine in mobile phase
injection 5 μL
detector 306 nm cell: 13 μL
Description
Featured Industry Food and Beverages
Legal Information SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany
suitability application for LC-MS