LC/MS Analysis of Sodium, Potassium and Chloride on SeQuant(R) ZIC-HILIC
LC/MS Analysis of Sodium, Potassium and Chloride on SeQuant® ZIC-HILIC
Conditions
column SeQuant ZIC-cHILIC, PEEK, 150 x 4.6 mm, 5 μm, 100 Ã… (1.50461.0001)
column temp. 30 °C
mobile phase [A] acetonitrile; [B] ammonium acetate 62.5mM pH 5; (80:20, A:B)
flow rate 1.0 mL/min
pressure 1088 psi 75 Bar
sample 100 μg/mL KCl and 100 μg/mL NaCl
injection 20 μL
detector Sedere Sedex 85 ELSD (cell: standard nebulizer)
Description
Featured Industry General Analytical
Legal Information SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany