Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS Analysis of Sucralose on SeQuant ZIC-HILIC
LC/MS Analysis of Sucralose on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-cHILIC, PEEK, 150 x 4.6 mm, 3 μm, 100Ã… (1.50661.0001)
column temp. 35 °C (oven), 35 °C (detector cell)
mobile phase [A] acetonitrile; [B] 25mM ammonium formate, adjusted with formic acid to pH 4.0; (90:10, A:B)
flow rate 0.7 mL/min
pressure 652 psi (45 bar)
sample Dissolve 20 mg of sucralose standard in 10 mL of diluent (Diluent: Mobile phase)
injection 20 μL
detector Refractive Index (RI) (cell: 10 μL)