Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS Analysis of Triglycerides in Corn Oil on Ascentis® C18
LC/MS Analysis of Triglycerides in Corn Oil on Ascentis® C18
Conditions
column two Ascentis C18, 15 cm x 2.1 mm I.D., 3 µm particles connected in series (581302-U)
column temp. 25 °C
mobile phase [A] 2-propanol: [B] acetonitrile; (60:40, A:B)
flow rate 0.2 mL/min
sample corn oil weighed and diluted in acetone (10 mg in 1 ml w/v)
injection 2 µL
detector APCI/MS positive ion mode
Description
Categories Analytical Chromatography, Fatty Acids, FAMEs
Featured Industry Food and Beverages