Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS Analysis of Vitamin D2, D3, Gamma- and Alpha-Tocopherol on Ascentis® Express C18
LC/MS Analysis of Vitamin D2, D3, Gamma- and Alpha-Tocopherol on Ascentis® Express C18
Conditions
column Ascentis Express C18, 10 cm x 2.1 mm I.D., 2.7 µm particles (53823-U)
column temp. 40 °C
mobile phase [A] water; [B] acetonitrile; (5:95, A:B)
flow rate 0.4 mL/min
sample 1 µg/mL in acetonitrile
injection 2 µL
detector MS, ESI+
Description
Analysis Note Analysis was conducted using standard solution, detection on LC/MS system.
Categories Analytical Chromatography, Clinical, vitamins, Vitamins and Natural products
Featured Industry Clinical
Vitamins, Nutraceuticals, and Natural Products