Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC-MS Anlaysis of Androsterone Isomers on Ascentis® Express Biphenyl
LC-MS Anlaysis of Androsterone Isomers on Ascentis® Express Biphenyl
Conditions
column Ascentis Express Biphenyl, 10cm x 2.1mm, 2.7mum particles (64065-U)
column temp. 35 °C
mobile phase [A] water with 0.1% formic acid; [B] acetonitrile with 0.1% formic acid 50:50 (A:B)
flow rate 0.2 mL/min
pressure 4200 psi (290 bar)
sample 500 ng/mL in mobile phase
injection 2 µL
detector MS, SIM, ESI+
Description
Analysis Note This application demonstrates the separation of Androsterone and trans-Androsterone on Ascentis Express Biphenyl. The shape selectivity of the biphenyl phase makes this a suitable column for analysis with baseline resolution.
Categories Analytical Chromatography, Steroids
Featured Industry Clinical
Pharmaceutical (small molecule)
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for LC-MS