Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC-MS Anlaysis of Pain Managment Opiates on Ascentis® Express Biphenyl, 30 x 2.1 mm
LC-MS Anlaysis of Pain Managment Opiates on Ascentis® Express Biphenyl, 30 x 2.1 mm
Conditions
column Ascentis Express Biphenyl, 3 cm x 2.1 mm, 2.7 µm particles (64054-U)
column temp. 30 °C
mobile phase [A] water with 0.1% formic acid; [B] acetonitrile with 0.1% formic acid;
gradient 10 to 20% B in 0.75 min; to 95% B in 1.5 min
flow rate 0.3 mL/min
pressure 2125 psi (147 bar)
sample 50-500 ng/mL in 99:1, water:methanol
injection 2 µL
detector MS-TOF, ESI+
Description
Analysis Note This application demonstrates the separation of 13 pain managment opiates on the Ascentis Express Biphenyl using an LC-TOF mass spectrometer. This application demonstrates the scalability of this method to 30 x 2.1 mm column geometry using the 2.7 um biphenyl phase. The method provides baseline resolution of all isobaric sets of interest.
Categories Analytical Chromatography, Drugs of Abuse, Pharmaceuticals, Opiate
Featured Industry Food and Beverages
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for LC-MS