LC/MS/MS Analysis of Pesticides in Cannabis on the Ascentis RP-Amide after QuEChERS Extraction and Cleanup Using Supel QuE Verde
LC/MS/MS Analysis of Pesticides in Cannabis on the Ascentis RP-Amide after QuEChERS Extraction and Cleanup Using Supel QuE Verde
Conditions
column Ascentis RP-Amide, 10 cm x 2.1 mm I.D., 3.0 µm
guard column Ascentis RP-Amide Supelguard guard cartridge, 2 cm x 2.1 mm I.D., 5 µm
mobile phase [A] 5 mM ammonium formate, 0.1% formic acid in 95:5 water:acetonitrile; [B] 5 mM ammonium formate, 0.1% formic acid in 5:95 water:acetonitrile
gradient 10% B held for 1 min; to 100% B in 13 min; held at 100% B for 6 min; to 10% in 0.5 min; held at 10% B for 6 min
flow rate 0.4 mL/min
column temp. 30 °C
detector MRM
injection 5 µL QuEChERS extract of cannabis in acetonitrile
Description
Featured Industry Food & Beverage
Legal Information Ascentis is a trademark of Sigma-Aldrich Co. LLC
Supel is a trademark of Sigma-Aldrich Co. LLC
Supelguard is a trademark of Sigma-Aldrich Co. LLC
Analysis Note In this application a majority of the LC-amenable pesticides on the Oregon list were extracted from cannabis spiked at 50 ng/g using QuEChERS, followed by cleanup with Supel QuE Verde. Analysis was achieved using the Ascentis RP-Amide column equipped with a guard.  Supel QuE Verde offered sufficient background reduction for LC/MS/MS analysis with recoveries of 70-120% for a majority of the pesticides.
Application No. ASC120118G