Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS/MS Analysis of Vitamins B1 and B6 on Ascentis® Express OH5
LC/MS/MS Analysis of Vitamins B1 and B6 on Ascentis® Express OH5
Conditions
column Ascentis Express  OH5, 10 cm x 2.1 mm I.D., 2.7 µm particles (53757-U)
column temp. 30 °C
mobile phase [A] water with 0.1% formic acid/10 mM ammonium formate; [B] acetonitrile with 0.1% formic acid/10 mM ammonium formate; (A:B, 15:85)
flow rate 0.2 mL/min
pressure 1150 psi (79 bar)
sample 10 ng/mL in mobile phase
injection 1 µL
detector MS, ESI(+), MRM mode
Description
Analysis Note The present work establishes an HILIC LC/MS/MS method for analysis of water soluble vitamins B1 and B6 on an Ascentis® Express OH5 column. LC/MS grade solvents were used to supply low background interference and low particulate contaminants for robust, trouble-free operation.
Categories Analytical Chromatography, Immunosuppressants
Featured Industry Vitamins, Nutraceuticals, and Natural Products