Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Loading Capacity of RNaseA on Proteomix® SCX-NP3
Loading Capacity of RNaseA on Proteomix® SCX-NP3
Conditions
column Proteomix SCX-NP3, 5 cm x 4.6 mm I.D., 3 µm particles (Z777151)
column temp. 25 °C
mobile phase 10 mM phosphate buffer, pH 6.0
gradient 0-1.0M NaCl (20 min)
flow rate 1.0 mL/min
injection 100 µL
detector UV, 280 nm
Description
Analysis Note Courtesy of Miyako Kawakatsu, M&S Instruments, Inc., This application note examines the loading capacity of Ribonuclease A on a Proteomix SCX-NP3 column. Sample sizes ranging from 50 μg to 5 mg were examined in this analysis.
Categories Analytical Chromatography, StiµLants
Featured Industry Life Science and Biopharma
Legal Information Proteomix is a registered trademark of Sepax Technologies