NARP-HPLC-APCI-MS Analysis of TAGs from Invernizzina Cheese on Ascentis(R) Express C18
NARP-HPLC-APCI-MS Analysis of TAGs from Invernizzina Cheese on Ascentis® Express C18
Conditions
column Ascentis Express C18, 15 cm x 4.6 mm I.D., 2.7 μm particles (53829-U)
mobile phase [A] acetonitrile; [B] 2-propanol (IPA)
gradient 0 to 70% B in 50 min; held at 70% B for 5 min; to 0% B in 1 min
flow rate 1 mL/min
sample lipid extract, 15 to 30 mg/mL in acetone
injection 7-20 μL
detector MS, APCI(+), mass range 250–1100 m/z
Description
Analysis Note Shown here is a procedure to extract lipids from cheese and analyse the TAG profile using LC/MS methodology.
Sample preparation: The lipid fraction was extracted by adding 10 g of the cheese sample (accurately weighed), 20 mL of 0.2 N hydrochloric acid, and 20 mL of ethyl alcohol to a volumetric flask. The flask was placed in a 50 °C water bath and gently shaken for 30 minutes with constant magnetic stirring until a homogenous suspension was formed. After cooling the flask to room temperature under running water, 210 mL of n-hexane and diethyl ether (1:2, v/v) were added and the mixture was shaken for an additional 15 minutes. The suspension was left to stand for 10 minutes to allow phase separation. This extraction protocol was repeated three times. The organic extracts were pooled, dried over anhydrous sodium sulfate, filtered, and taken to dryness under vacuum. The final dry residue was stored at -18 °C until use. Samples ranging from 15 to 30 mg of each of the dry residue from the final lipid extracts were diluted with 1 mL of acetone and filtered through a 0.45 μm Acrodisc nylon membrane filter prior to LC/MS analysis.
Featured Industry Food and Beverages
Life Science and Biopharma
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Shimadzu LCMS-2010 APCI(+): range 250-1100 m/z; nebulizing gas (N2) flow: 2.0 L/min; heat block: 300 °C, CDL temperature: 300 °C; interface temperature: 450 °C; detector voltage: 1.6 kV, Shimadzu LCMS-2010 APCI(+): range 250-1100 m/z;
nebulizing gas (N2) flow: 2.0 L/min;
heat block: 300 °C, CDL temperature: 300 °C;
interface temperature: 450 °C;
detector voltage: 1.6 kV