Retention Time Stability after Repeated Injections on SeQuant® ZIC-HILIC
Retention Time Stability after Repeated Injections on SeQuant® ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 50 x 4.6 mm, 5 μm, 200 Å (1.50451.0001)
mobile phase [A] acetonitrile; [B] 5 mM ammonium acetate; (80:20, A,B)
flow rate 0.5 mL/min
pressure 158 psi (1.1 MPa)
column temp. 24 °C
detector 254 nm
injection: 5 μL
Description
Featured Industry General Analytical
Legal Information SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany