Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Scalability of Separations on SeQuant® ZIC-HILIC. Analytical to Semi-Preparative Scale
Scalability of Separations on SeQuant® ZIC-HILIC. Analytical to Semi-Preparative Scale
Conditions
column SeQuant ZIC-HILIC, PEEK, 50, x 2.1 mm, 5 μm, 200 Å (1.50450.0001,1.50451.0001)
mobile phase [A] acetonitrile; [B] 5 mM ammonium acetate; (80:20, A:B)
flow rate 0.1, 0.5, 1.3, 9.3 mL/min
pressure 144, 144, 115, 158 psi (1.0, 1.0, 0.8, 1.1 MPa)
detector 254 nm
injection: 1, 5, 20, 100 μL
Description
Featured Industry General Analytical
Legal Information SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany