UHPLC Analysis of Benzoyl Peroxide from Clean & Clear® Continuous Control Acne Cleanser on Purospher® STAR RP18e Hibar® HR
UHPLC Analysis of Benzoyl Peroxide from Clean & Clear® Continuous Control Acne Cleanser on Purospher® STAR RP18e Hibar® HR
Conditions
sample preparation Clean & Clear® continuous control acne cleanser at 100 mg/mL in 30:70 acetonitrile: 5 mM potassium phosphate pH 3.0 in water. (See "Sample Prep" at Description Text)
column Purospher STAR RP18e Hibar HR, 10 cm x 2.1 mm I.D., 2.0 μm, 120Å (1.50648.0001)
mobile phase [A] 5 mM potassium phosphate, pH 3.0 with phosphoric acid in water; [B] acetonitrile
gradient 40 to 80% B in 9 min; hold at 80% B for 2 min; to 40% B in 0.1 min; to hold at 40% B for 6.9 min
flow rate 0.3 mL/min
pressure 4240 psi (292 bar)
column temp. 30 °C
Description
Categories Analytical/Chromatography, Antiinflammatories, Applications, HPLC Applications
Analysis Note Benzoyl peroxide is split into benzoic acid and oxygen when it comes in contact with the skin breaking down the keratin in the outermost layer. This helps to lower bacterial populations in the pores on the skin. It also has a mild drying effect, which allows excess oils and dirt to be easily washed away from the skin. This application demonstrates reverse phase interactions with an improvement on the current monograph.
Sample prep: 1 gram of Clean & Clear® continuous control acne cleanser (10% benzoyl peroxide) was added to a 25 mL volumetric flask and 10 mL of acetonitrile was added by pipet. The gel and acetonitrile was vortexed for 30 seconds and sonicated for one hour. 1mL of the acetonitrile was then filted with a 0.45 μm Millex®LH filter. The filtered acetonitrile (10 μL) was added to 9.99 mL of 30:70 acetonitrile:5 mM potassium phosphate (pH 3.0) in water with phosphoric acid to give a concentration of 100 mg/mL.
Legal Information Clean & Clear is a registered trademark of Johnson & Johnson
Hibar is a registered trademark of Merck KGaA, Darmstadt, Germany
Millex is a registered trademark of Merck KGaA, Darmstadt, Germany
Purospher is a registered trademark of Merck KGaA, Darmstadt, Germany