Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
UHPLC Analysis of Benzoyl peroxide from Neutrogena(R) Rapid Clear(R) stubborn acne spot gel on Purospher(R) STAR RP-18e Hibar(R) HR
UHPLC Analysis of Benzoyl peroxide from Neutrogena® Rapid Clear® stubborn acne spot gel on Purospher® STAR RP-18e Hibar® HR
Conditions
sample preparation 1 gram of Neutrogena® Rapid Clear® Stubborn Acne Spot Gel (10% benzoyl peroxide). (See "Sample Prep" at Description Text.)
column Purospher STAR RP-18e Hibar HR, 10 cm x 2.1 mm I.D., 2.0 μm, 120Ã… (1.50648.0001)
column temp. 30 °C ambient
mobile phase [A] 5 mM Potassium Phosphate, pH 3.0 with phosphoric acid in water; [B] Acetonitrile
gradient 40 to 80% B in 9 min; hold at 80% B for 2 min; to 40% B in 0.1 min; to hold at 40% B for 6.9 min
flow rate 0.3 mL/min
pressure 4236 psi (292 bar)
sample 100 μg/mL Benzoyl Peroxide in 30:70 acetonitrile:5mM potassium phosphate pH 3.0 in water with phosphoric acid
injection 1 μL
detector UV, 235 nm
Description
Analysis Note Benzoyl peroxide is split into benzoic acid and oxygen when it gets in contact with the skin breaking down the keratin in the outermost layer. This helps to lower bacterial populations in the pores on the skin. It also has a mild drying effect, which allows excess oils and dirt to be easily washed away from the skin. This application demonstrates reverse phase interactions with an improvement on the current monograph.
Sample prep: 1 gram of Neutrogena® Rapid Clear® Stubborn Acne Spot Gel (10% benzoyl peroxide) was added to a 25mL volumetric flask and 10mL of acetonitrile was added by pipet. The gel and acetonitrile was vortexed for 30 seconds and sonicated for one hour. 1mL of the acetonitrile was then filled with a 0.45 μm Millex®-LH filter. 10μL of the filtered solution was added to 9.99mL of 30:70 acetonitrile:5mM potassium phosphate pH 3.0 in water with phosphoric acid to give a concentration of 100 mg/mL.
Featured Industry Cosmetics, Personal Care, and Cleaning Products
Legal Information Neutrogena is a registered trademark of Johnson & Johnson, Purospher is a registered trademark of Merck KGaA, Darmstadt, Germany, Millex is a registered trademark of Merck KGaA, Darmstadt, Germany, Rapid Clear is a registered trademark of Johnson & Johnson, Hibar is a registered trademark of Merck KGaA, Darmstadt, Germany
suitability application for UHPLC