UHPLC Analysis of Bupropion and Five Related Compounds on Ascentis® Express C18
Conditions
instrument Dionex/Thermo Electron Ultimate 3000 UHPLC 
column Ascentis® Express C18, 10 cm x 3.0 mm, 2.7 µm (53814-U)
mobile phase   [A] 25 mM potassium phosphate buffer in water, pH 2.5 with phosphoric acid [B] acetonitrile gradient: 20% B for 1 min, then 20% to 35% B in 6 min; to 20% B in 0.1 min; hold at 20% B for 1.4 min
gradient 20% B for 1 min, then 20% to 35% B in 6 min; to 20% ] in 0.1 min; hold at 20% B for 1.4 min
flow rate  0.6 mL/min
pressure 3000 psi
column temp. 35 °C
detector UV, 200 nm 
injection 5.0 µL
Description
Featured Industry Pharmaceutical (Small Molecule)
Legal Information Ascentis is a trademark of Sigma-Aldrich Co. LLC
Dionex and Ultimare are trademarks of Thermo Fisher Scientific Inc
Analysis Note Bupropion is primarily a medication used as an antidepressant or smoking cessation aid. Bupropion acts as a norepinephrine-dopamine reuptake inhibitor (NDRI) and is also a nicotinic antagonist. These standards are used in testing methods by HPLC and LC/MS for applications in pharmaceutical research, forensic analysis and clinical toxicology.  Shown here is a gradient separation of bupropion and five of its related compounds in less than ten minutes on an Ascentis Express C18 column. Using bupropion CRM provides reliable identification and quantification.