Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
UHPLC Analysis of Human IgA on BIOshell(TM) IgG 1000 Ã… C4
UHPLC Analysis of Human IgA on BIOshell™ IgG 1000 Ã… C4
Conditions
column BIOshell IgG 1000 Ã… C4; 10 cm x 2.1 mm I.D., 2.7 μm particles (63288-U)
column temp. 75 °C
mobile phase [A] 0.1% TFA (water); [B] 0.1% TFA (acetonitrile)
gradient 30% B to 60% B in 10 min; 60% B to 100% B in 10 min
flow rate 0.2 mL/min
pressure 1425 psi (98 bar)
sample Human IgA, 100 μg/mL, 0.1% TFA (water)
injection 1.0 μL
detector UV, 215 nm
Description
Analysis Note Human immunoglobulin A (IgA) can exist in two forms: a monomer (~150 kDa) in the serum and a dimer (~300 kDa) in mucosal secretory tissues. Serum IgA (monomeric) functions as an anti-inflammatory antibody by interacting with CD89 on immune effector cells. Serum IgA was chromatographically analyzed on a BIOshell IgG 1000 Å C4 column and compared to a fully porous, 300 Å C4 column. The BIOshell IgG column displayed superior resolution and recovery over the competitor column.
Featured Industry Life Science and Biopharma
Legal Information BIOshell is a trademark of Sigma-Aldrich Co. LLC
suitability application for UHPLC