UHPLC Analysis of Intact Infliximab on BIOshell(TM) IgG 1000 Ã… C4
UHPLC Analysis of Intact Infliximab on BIOshell™ IgG 1000 Ã… C4
Conditions
column BIOshell IgG 1000 Ã… C4; 10 cm x 2.1 mm I.D., 2.7 μm particles (63288-U)
column temp. 75 °C
mobile phase [A] 70:25:5, water (0.1% TFA): acetonitrile (0.1% TFA): 1-butanol (0.1% TFA); [B] 50:45:5, water (0.1% TFA): acetonitrile (0.1% TFA): 1-butanol (0.1% TFA)
gradient 0% B to 100% B in 15 min
flow rate 0.2 mL/min
pressure 1800 psi (122 bar)
sample Infliximab, 100 μg/mL, water (0.05% TFA)
injection 5 μL
detector UV, 215 nm
Description
Analysis Note Infliximab (brand name Remicade) is an immunosuppressive biologic drug used to treat rheumatoid arthritis, psoriatic arthritis, Crohn′s disease, and many other diseases by blocking TNF alpha which prevents an autoimmune reaction. It has a molecular weight of ~150 kDa. The BIOshell IgG 1000 Ã… C4 UHPLC column yielded an efficient peak for this protein in less than five minutes.
Featured Industry Pharmaceutical (small molecule)
Legal Information BIOshell is a trademark of Sigma-Aldrich Co. LLC
suitability application for UHPLC