Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
UHPLC Analysis of Turmeric Curcumin Herbal Supplement on Ascentis® Express RP-Amide
UHPLC Analysis of Turmeric Curcumin Herbal Supplement on Ascentis® Express RP-Amide
Conditions
sample preparation Filter with 0.22 µm PVDF
sample preparation Sonicate 20 min
extraction Weigh approximately 100 mg of herbal supplement ad add 100 mL ethanol
column Ascentis Express RP-Amide, 10 cm x 4.6 mm I.D., 5 µm particles (50774-U)
column temp. 35 °C
mobile phase [A] water with 0.1% formic acid; [B] acetonitrile with 0.1% formic acid;
gradient 40% B held for 1 min; 40 to 100% B in (6 min); 100% B held for 5 min
flow rate 1.0 mL/min
pressure 1270 psi (88 bar)
sample turmeric curcumin hernal supplement extract in ethanol dilute 1:1 with water
injection 5 µL
detector UV, 250 nm
Description
Categories Analytical Chromatography, Anti-inflammatories, Antioxidants
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for UHPLC