Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
UHPLC Gradient Analysis of Boswellia Herbal Supplement on Ascentis® Express Phenyl-Hexyl
UHPLC Gradient Analysis of Boswellia Herbal Supplement on Ascentis® Express Phenyl-Hexyl
Conditions
column Ascentis Express Phenyl-Hexyl, 10 cm x 3.0 mm I.D., 2.7 µm particles (53345-U)
column temp. 35 °C
mobile phase [A] 0.1% phosphoric acid; [B] acetonitrile
gradient 50% B for 0.5 min; to 100% B in 4.5 min; held at 100% B for 5 min
gradient 0.6 mL/min
pressure 2390 psi (165 bar)
injection 2 µL
detector UV, 210 nm
Description
Application extraction:
  1. Weigh 100 mg Bosewellia Herbal Supplement
    2. Add 10 mL ethanol
    3. Sonicate 30 min at 40°C
    4. Dilute 100 μL extract, 400 μL ethanol, 500 μL water
Categories Analytical Chromatography, Herbal medicines
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for UHPLC