Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
UHPLC/MS Analysis of Estradiols using Ascentis® Express C18
UHPLC/MS Analysis of Estradiols using Ascentis® Express C18
Conditions
column Ascentis Express C18, 10 cm x 2.1mm, 2 µm particles (50813-U)
column temp. 30 °C
mobile phase [A] water; [B] methanol
gradient 30 to 40% B in 0.1 min; 40 to 60% B in 0.9 min; 60 to 75% B in 6 min; 75 to 30% B in 0.1 min; held at 30% B for 2.9 min
flow rate 0.2 mL/min
pressure 6544 psi (451 bar)
sample in 20% methanol -80% water at 10 ng/mL
injection 10 µL
detector MS, ESI (-), 271/145
Description
Analysis Note Standard at 10 ng/mL.
Featured Industry Food and Beverages
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for UHPLC
Materials
Related Links
Categories